Learn More
Enzo Life Sciences ORGANELLE-ID-RGB reagent IV (200µl)
SDP
Supplier:  Enzo Life Sciences ENZ53009C200
Encompass_Preferred
The ORGANELLE-ID-RGB Reagent IV is a mixture of cell-permeable red fluorescent endoplasmic reticulum dye, green fluorescent lysosomal dye and blue fluorescent nucleic acid dye. The staining pattern arising from the combination of these three dyes permits visualization of the target organelles by fluorescence/confocal microscopy. The reagent, supplied as a 500X solution, is sufficient for 1000 microscopy assays. The single-tube format makes this multi-organelle stain reagent easy to use. Purity: ≥93% (HPLC). Long Term Storage: -80°C.
Catalog No. 50-201-2654