Learn More
Perkin Elmer US LLC Organics Burner Drain Assembly for PinAAcle and AAnalyst
Supplier:  Perkin Elmer US LLC N3160683
Encompass_Preferred
Note that this kit contains the drain assembly and bottle only and does not include the burner assembly or any burner components.
Catalog No. NC1984926