Learn More
Strem, An Ascensus Company CAS 335-36-4. 25g. Perfluoro-2-butyltetrahydrofuran min. 95%. MFCD00014513
SDP
Supplier:  Strem, An Ascensus Company 09591025G
Encompass
CAS 335-36-4. 25g. Perfluoro-2-butyltetrahydrofuran min. 95%. MFCD00014513. Molecular Weight 416.09. Molecular Formula C8F16O. Color/form colorless liq. Strem 09-5910. http//www.strem.com/catalog/v/09-5910/
Catalog No. 50-211-0705
 Restricted Item