Learn More
Selleck Chemical LLC ORIC-944 E1967-25MG
SDP
Supplier:  Selleck Chemical LLC E196725MG
Encompass_Preferred
ORIC-944 is a potent and selective allosteric inhibitor of the polycomb repressive complex 2 (PRC2) via the embryonic ectoderm development (EED) subunit It exhibits anti-tumor activity in vivo prostate cancer models
Catalog No. 50-000-39132