Learn More
Grainger O-Rings and O-Ring Kits 5/PK
Supplier:  Grainger 713R22
Encompass_Preferred
O-rings are circular seals used in machinery and equipment to create a tight seal between stationary and moving parts. They are available in various materials like rubber, silicone, PTFE, neoprene, nitrile, EPDM, and more. O-rings come in standard and custom sizes. O-ring kits include assortments of O-rings in a range of sizes.
Catalog No. NC2882471