Learn More
Selleck Chemical LLC Ornidazole E6017-1G
SDP
Supplier:  Selleck Chemical LLC E60171G
Encompass_Preferred
Ornidazole (Levo-) (Levornidazole (S)-Ornidazole) the levo-isomer of ornidazole is a third-generation nitroimidazole derivative It exhibits antiprotozoal and antibacterial properties against anaerobic bacteria
Catalog No. 50-000-41708