Learn More
ATCC Oropouche Virus Immune Ascitic Fluid, Freeze-dried (ATCC VR-1228AF, V-505-701-562)
Supplier: ATCC VR1228AF
Oropouche virus immune ascitic fluid is a freeze-dried mouse hyperimmune ascitic fluid containing polyclonal antibodies against Oropouche virus, supplied for laboratory research use. It is intended for immunostaining and antibody-based detection of Oropouche virus antigens; it is not for diagnostic, therapeutic, or human/animal consumption.
- Freeze-dried ascitic fluid
- Contains polyclonal anti-Oropouche virus antibodies
- Intended for research use only
- Suitable for immunostaining and antigen detection
- Biosafety level: BSL 1
- Storage: not listed on product page
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.