Learn More
TARGETMOL CHEMICALS INC Orotic acid zinc 1G
SDP
Supplier:  TARGETMOL CHEMICALS INC T32711G
Encompass_Preferred
Also available in 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. Orotic acid zinc (Zinc Orotate) salt dihydrate is an intermediate product in pyrimidine synthesis. Purity 100%
Catalog No. 50-287-7851