Learn More
TARGETMOL CHEMICALS INC oroxylin A 7-O-beta-d-glucuron
SDP
Supplier:  TARGETMOL CHEMICALS INC TN67821MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Oroxylin A 7-O-beta-d-glucuronide was a new alkaloid which found in the bulbs of Lycoris radiata. Purity 99.02%
Catalog No. 50-287-9628