Learn More
Medchemexpress LLC HY-N2481 5mg Medchemexpress, Oroxylin A-7-O-glucuronide CAS:36948-76-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN24815MG
Encompass_Preferred
Medchemexpress, HY-N2481 5mg Oroxylin A-7-O-glucuronide CAS:36948-76-2 Oroxylin A-7-O-glucuronide (Oroxyloside; Oroxylin A-7-O-β-D-glucuronide) is a flavonoid glucuronide isolated from the dried root of Scutellaria baicalensis, with prolyl oligopeptidase (POP) inhibitory activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2898