Learn More
Selleck Chemical LLC Orphenadrine Hydrochloride
SDP
Supplier:  Selleck Chemical LLC S566425MG
Encompass_Preferred
Orphenadrine Hydrochloride is an uncompetitive NMDAR antagonist H1 receptor antagonist and nonselective mAChR antagonist with muscle relaxant activity
Catalog No. 50-000-03424