Learn More
Cayman Chemical OrphenadrInhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 2639110 MG
Encompass_Preferred
A muscarinic acetylcholine receptor antagonist (Kds = 48, 213, 120, 170, and 129 nM for M1-M5 mAChRs, respectively); decreases oxotremorine-induced muscle activity in rabbits at 2-5 mg/kg; an NMDA receptor antagonist (Ki = 6 µM in human postmortem frontal cortex; IC50 = 16.2 µM in cultured superior colliculus neurons)
Catalog No. 50-293-3932