Learn More
Medchemexpress LLC Orsellinic acid | 480-64-8 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN31265MG
Encompass_Preferred
Orsellinic acid is a benzoic acid derivative that can be isolated from *Chaetomium globosum* endophytic on *Ephedra fasciculata* (Mormon tea). It blocks PAF-mediated apoptosis, inhibits caspase-3/7 activation, and PARP cleavage. This compound is utilized in research concerning neurons and various tumors, including non-small cell lung cancer, breast cancer, neuroblastoma, and pancreatic cancer.
  • Blocks PAF-mediated apoptosis
  • Inhibits caspase-3/7 activation
  • Inhibits PARP cleavage
  • Utilized in research concerning neurons
  • Utilized in research concerning various tumors
Catalog No. 50-004-21310