Learn More
Med Vet International OrthopedicTwist Drill Bit, 5/64", 2.0mm, J0372B, Each
Supplier: Med Vet International JOR372B
Orthopedic Drill Bit. Jorvet drill bits are European-made. Razor sharp and stainless steel 5". 5/64" 2.0mm 2mm Sherman Self-Tapping System. The 2mm self-tapping system enables the veterinary surgeon to plate the bones of cats and miniature dogs effectivel