Learn More
Medchemexpress LLC Orvepitant maleate | 579475-24-4 | 100.0% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY122347A10MG
Encompass_Preferred
Orvepitant maleate is the maleate salt of orvepitant, a potent and selective neurokinin-1 (NK-1) receptor antagonist used as a research compound for studying NK-1 receptor pharmacology and related central nervous system indications.
  • Potent, selective NK-1 receptor antagonist suitable for in-vitro and in-vivo studies.
  • Maleate salt with molecular weight 744.70 g/mol.
  • High chemical purity reported by the manufacturer.
  • Soluble in DMSO for preparing concentrated stock solutions.
  • Used in research on depressive disorders and chronic refractory cough models.
Catalog No. 50-002-31247