Learn More
Medchemexpress LLC Oryzalin | 19044-88-3 | 99.6% | 346.36 | 100 MG
Supplier: Medchemexpress LLC HY147092100MG
Oryzalin is a dinitroaniline herbicide that exhibits antimitotic and antileishmanial activities. It functions by binding to plant tubulin, thereby inhibiting the polymerization of microtubules in plant cells and affecting chromosome migration. It can also be employed to induce chromosome doubling.
- Functions as a dinitroaniline herbicide
- Exhibits antimitotic and antileishmanial activities
- Binds to plant tubulin
- Inhibits microtubule polymerization in plant cells
- Affects chromosome migration
- Can be used to induce chromosome doubling