Learn More
Medchemexpress LLC Oryzalin | 19044-88-3 | 99.6% | 346.36 | 50 MG
Supplier: Medchemexpress LLC HY14709250MG
Oryzalin is a dinitroaniline herbicide that exhibits antimitotic and antileishmanial properties. It works by binding to plant tubulin, which inhibits the polymerization of microtubules in plant cells and affects chromosome migration. Oryzalin can also be utilized to induce chromosome doubling. It is intended for research use only and is not sold to patients.
- It acts as a dinitroaniline herbicide.
- It exhibits antimitotic activity.
- It demonstrates antileishmanial activities.
- It can bind to plant tubulin.
- It inhibits the polymerization of microtubules in plant cells.
- It affects chromosome migration.
- It can induce chromosome doubling.