Learn More
MSC Rubbermaid Liquid Dispenser, Counter Mount, Touch-Free, Polished Chrome, ADA Compliant
Supplier:  MSC 57845927
Encompass
The One Shot Liquid Dispenser in Polished Chrome offers automatic, touch-free dispensing of concentrated soaps, promoting healthy hand washing and reducing waste. Its reliable counter-mounted design utilizes Smart Sensor™ technology for precise delivery and helps earn LEED credits.
  • Eliminates overuse and costly soap waste.
  • Reduces spread of germs through touch-free operation.
  • High yield: up to 2,000 hand washes per refill.
  • Disposable hygienic refill for fresh, germ-free soap.
  • Minimizes waste with industry-leading evacuation.
  • Designed to prevent clogs, leaks, breakdowns, and mess.
  • Reliable, low-maintenance dispensing.
  • Environmental choice: recyclable refills, biodegradable soap, Green Seal™ certified soaps, up to 80% packaging waste reduction.
  • Long battery life: up to 3 years or 120,000 cycles for liquid dispensers.
Catalog No. 50-283-6807