Learn More
Cayman Chemical OseltamIvIr AcId 5mg
SDP
Supplier:  Cayman Chemical 157795 MG
Encompass_Preferred
The active metabolite of the antiviral prodrug oseltamivir; acts as an inhibitor of influenza neuraminidases A and B (IC50 values are 0.1 to 4.9 nM for both enzymes), in this way preventing virus budding and release
Catalog No. 50-297-0766