Learn More
Medchemexpress LLC Oseltamivir acid (hydrochloride) | 1415963-60-8 | >98.0% | C14H25ClN2O4 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13318A10MG
Encompass_Preferred
Oseltamivir acid hydrochloride is the active metabolite of the antiviral prodrug oseltamivir and acts as a potent neuraminidase inhibitor. Supplied as a solid hydrochloride salt for research use, it is suitable for enzymatic, biochemical, and cell-based antiviral studies.
  • Active metabolite of oseltamivir.
  • Potent neuraminidase inhibitor.
  • Supplied as a solid hydrochloride salt.
  • High purity (≥98.0%).
  • Suitable for enzymatic and cell-based assays.
  • Available in small research pack sizes (mg scale).
Catalog No. 50-002-60247