Learn More
Medchemexpress LLC HY-17016 500mg , Oseltamivir (phosphate) GS 4104 CAS:204255-11-8 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY17016500MG
Encompass_Preferred
Medchemexpress, HY-17016 500mg Oseltamivir (phosphate) GS 4104 CAS:204255-11-8 Formula:C16H31N2O8P Influenza A and B Purity:98% Oseltamivir phosphate is a neuraminidase inhibitor recommended for the treatment and prophylaxis of influenza A and B . Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2432