Learn More
Apexbio Technology LLC Oseltamivir phosphate 204255-11-8 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC B18035.1
Encompass_Preferred
Oseltamivir phosphate known pharmacologically as GS-4104 functions as a selective competitive inhibitor targeting neuraminidase an enzyme essential for influenza viral particle release from infected cells By inhibiting neuraminidase activity this compound restricts influenza viral propagation within host cells Oseltamivir phosphate demonstrates inhibitory activity against influenza A and B viruses with reported neuraminidase inhibitory potency (IC50) typically in the low nanomolar range ( 1 5 nM) In biomedical research it is primarily utilized to evaluate antiviral efficacy investigate neuraminidase-inhibition mechanisms and facilitate influenza infection modeling in preclinical experimental systems and drug screening assays
Catalog No. 50-000-62349