Learn More
Selleck Chemical LLC Oseltamivir Phosphate 250mg 204255-11-8
SDP
Supplier:  Selleck Chemical LLC S2597250MG
Encompass_Preferred
Oseltamivir Phosphate is a potent and selective inhibitor of the neuraminidase that is essential for replication of influenza A and B viruses, used to prevent influenza. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-2610