Learn More
MSC OSG Thread Limit Gage, M24 x 3.00, 6H Class, Go/No-Go
Supplier: MSC 55440069
This metric plug thread gage is used to inspect the pitch diameter and pitch accuracy of internal threads. It is a double-end gage, featuring both Go and No-Go ends, and comes with a handle. Manufactured to Class X tolerances per ANSI B1.16M, these gages are ground from premium high-speed steel (HSS) and hardened to 64Rc to ensure superior wear resistance.
- Available with short form or long form certification
- Available in standard 6H class of fit
- Hardened to 64 HrC for superior wear resistance
- Made from premium high-speed steel
- Includes a handle