Learn More
Selleck Chemical LLC OSI-027 S2624-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S262410MM/1ML
Encompass_Preferred
OSI-027 (ASP4786 CERC 006 AEVI-006) is a selective and potent dual inhibitor of mTORC1 and mTORC2 with IC50 of 22 nM and 65 nM in cell-free assays and more than 100-fold selectivity observed for mTOR than PI3K PI3K PI3K or DNA-PK OSI-027 induces autophagy in cancer cells
Catalog No. 50-311-2508