Learn More
Selleck Chemical LLC OSI-027 50mg 936890-98-1 ASP4786
SDP
Supplier:  Selleck Chemical LLC S262450MG
Encompass_Preferred
OSI-027 (ASP4786) is a selective and potent dual inhibitor of mTORC1 and mTORC2 with IC50 of 22 nM and 65 nM in cell-free assays, and more than 100-fold selectivity observed for mTOR than PI3K?, PI3K?, PI3K? or DNA-PK. OSI-027 induces autophagy in cancer cells. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-878-4