Learn More
TARGETMOL CHEMICALS INC OSILODROSTAT 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T427710MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Osilodrostat (LCI699) (LCI699) is an effective inhibitor of human 11(beta)-hydroxylase (IC50 2.5 nM) and aldosterone synthase (IC50 0.7 nM). purity: 99%
Catalog No. 50-257-6782