Learn More
Medchemexpress LLC Osilodrostat (phosphate) | 1315449-72-9 | 98.8% | 325.23 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY16276A
Encompass_Preferred
Osilodrostat phosphate is a potent, orally active 11β-hydroxylase (CYP11B1) inhibitor with an IC50 value of 35 nM. It also acts as a potent, orally active aldosterone synthase (CYP11B2) inhibitor. This compound inhibits aldosterone and corticosterone synthesis and has blood pressure-lowering ability, making it suitable for research related to Cushing syndrome (CS).
  • Inhibits cortisol and aldosterone in various cell types.
  • Reduces mean arterial pressure in animal models.
  • Used in research for Cushing syndrome.
Catalog No. 50-003-95427