Learn More
Medchemexpress LLC HY-15772 200mg , Osimertinib CAS:1421373-65-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY15772200MG
Encompass_Preferred
HY-15772 200mg Osimertinib CAS: 1421373-65-0 Osimertinib (AZD9291) is a covalent, orally active, irreversible, and mutant-selective EGFR inhibitor with an apparent IC50 of 12 nM against L858R and 1 nM against L858R/T790M, respectively. Osimertinib overcomes T790M-mediated resistance to EGFR inhibitors in lung cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0175