Learn More
Selleck Chemical LLC Osimertinib
SDP
Supplier:  Selleck Chemical LLC S72971G
Encompass_Preferred
Osimertinib (AZD9291) is an oral irreversible and mutant-selective EGFR inhibitor with IC50 of 12 92 11 44 and 493 8 nM for Exon 19 deletion EGFR L858R/T790M EGFR and WT EGFR in LoVo cells respectively Phase 3
Catalog No. 50-000-04387