Learn More
Medchemexpress LLC Osimertinib dimesylate | 2070014-82-1 | 99.19% | C30H41N7O8S | 1 ML

Supplier: Medchemexpress LLC HY7907710MM/1ML
Osimertinib dimesylate is an irreversible and mutant selective EGFR inhibitor with IC50s of 12 and 1 nM against EGFRL858R and EGFRL858R/T790M, respectively. This product is intended for research use only.
- Purity: 99.19%
- CAS number: 2070014-82-1
- Solubility: 10 mM * 1 mL in water (ready for reconstitution)
- Data sheet, certificate of analysis, safety data sheet, and handling instructions are available
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.