Learn More
Medchemexpress LLC Osimertinib dimesylate | 2070014-82-1 | 99.19% | C30H41N7O8S2 | 200 MG
SDP
Supplier:  Medchemexpress LLC HY79077200MG
Encompass_Preferred
Osimertinib dimesylate (AZD-9291 dimesylate) is an irreversible and mutant-selective EGFR inhibitor. It demonstrates potent inhibitory activity against key EGFR mutations, including T790M, with high efficacy in tumor models. This product is for research use only.
  • Mutant selective EGFR inhibitor
  • Irreversible binding action
  • Potent inhibition against EGFR mutations
  • Improved pharmacokinetic profile
  • Broad chemical and profile diversity
  • Proven efficacy in tumor models
Catalog No. 50-004-34871