Learn More
Medchemexpress LLC Osimertinib dimesylate | 2070014-82-1 | 99.2% | 10 MG

Supplier: Medchemexpress LLC HY7907710MG
Osimertinib dimesylate is an irreversible, mutant-selective epidermal growth factor receptor (EGFR) inhibitor supplied as a solid research standard for in vitro and analytical applications. It demonstrates potent activity against activating and resistance EGFR mutants, with reported IC50 values of 12 nM (EGFR L858R) and 1 nM (EGFR L858R/T790M).
- Irreversible, mutant-selective EGFR inhibitor for research and analytical use.
- Demonstrated potency: IC50 12 nM (EGFR L858R) and 1 nM (EGFR L858R/T790M).
- High purity (99.19%) suitable for biochemical assays and analytical standards.
- Solid powder form, available in small mass sizes such as 10 mg.
- Chemical formula C30H41N7O8S2; molecular weight 691.82 g/mol.
- Storage: store sealed away from moisture; in solvent -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.