Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC OSM Human 209a.a 2ug
SDP

Supplier:  Medchemexpress LLC HYP7052A2UG

Encompass_Preferred

Recombinant human Oncostatin M (OSM) is a 209-amino-acid cytokine of the IL-6 family involved in hematopoiesis, mesenchymal stem cell differentiation, liver regeneration, heart remodeling, nociception, inflammation, and metabolism. Supplied lyophilized and tag-free, this recombinant protein is provided for in vitro research applications requiring biologically active OSM.

  • Contains 209 amino acids.
  • Expressed in E. coli and tag free.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Approximate molecular weight 24-26 kDa under reducing conditions.
  • Lyophilized from PBS (pH 7.4) with 8% trehalose for stability.
  • Do not reconstitute below 100 μg/mL in ddH2O.
  • Store lyophilized at -20°C; freeze aliquots at -20°C or -80°C for extended storage.

Catalog No. 50-001-85153


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.