Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC OSM Human 227a.a 100ug
SDP

Supplier:  Medchemexpress LLC HYP7052100UG

Encompass_Preferred

Recombinant human Oncostatin M (OSM) is a 227-amino-acid IL-6 family cytokine involved in hematopoiesis, tissue regeneration, inflammation, and metabolism. The product is expressed in E. coli, supplied tag-free and lyophilized after dialysis into PBS with trehalose, and shows an approximate molecular weight of 25.9 kDa by SDS-PAGE. It is intended for in vitro research applications.

  • Contains full-length human OSM sequence (227 aa).
  • Tag-free protein for native activity studies.
  • Expressed in E. coli and lyophilized for stability.
  • Formulated in PBS with 8% trehalose to protect during storage.
  • Approximate molecular weight 25.9 kDa (SDS-PAGE).
  • Recommended storage at -20°C; freeze aliquots for long-term storage.

Catalog No. 50-001-84834


May include imposed supplier surcharges.
Only null left
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.