Learn More
Medchemexpress LLC Osmi-1 | 1681056-61-0 | 99.9% | 500 MG

Supplier: Medchemexpress LLC HY119738500MG
OSMI-1 is a cell-permeable small-molecule inhibitor of O-GlcNAc transferase (OGT) used to reduce protein O-GlcNAcylation in mammalian cell studies. Supplied as a solid (white to light yellow), it is provided in multiple sizes and has defined storage recommendations for powder and solutions.
- Cell-permeable O-GlcNAc transferase inhibitor with reported IC50 of 2.7 μM.
- High purity: 99.9% by HPLC.
- Molecular weight 563.64 Da; formula C28H25N3O6S2.
- Solid appearance, white to light yellow.
- Available in milligram to gram quantities for research use.
- Recommended storage: powder at -20°C for long term or in solvent at -80°C for long term.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.