Learn More
Selleck Chemical LLC OSMI-1-5mg
SDP
Supplier:  Selleck Chemical LLC S98355MG
Encompass_Preferred
OSMI-1 is a cell-permeable O-GlcNAc transferase (OGT) inhibitor with IC50 of 2.7 μM for full length human OGT (ncOGT).
Catalog No. 50-238-9881