Learn More
Abcam OSMI-1, O-GlcNAc transferase (OGT) inhibitor, 5MG
Supplier:  Abcam AB2354555MG
Encompass_Preferred
MW 563.6 Da, Purity >98%. Potent, cell-permeable, O-GlcNAc transferase (OGT) inhibitor. Inhibits protein O-GlcNAcylation in several mammalian cell lines without qualitatively altering cell surface N- or O-linked glycans.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-6526