Learn More
Medchemexpress LLC OSMI-4 100mg | 2260791-14-6 | 100 MG

Supplier: Medchemexpress LLC HY114361100MG
OSMI-4 is a small-molecule inhibitor of O-GlcNAc transferase (OGT) used for biochemical and cellular research. It shows cellular activity with an EC50 of approximately 3 μM and is supplied as a purified solid for experimental assays.
- Cellular EC50 ~3 μM against O-GlcNAc transferase.
- High purity, 99.7%.
- Molecular formula C27H26ClN3O7S2; molecular weight 604.09 g·mol⁻¹.
- Available as a 100 MG solid pack.
- Recommended storage: powder -20 °C (up to 3 years) or 4 °C (up to 2 years).
- Structural identifier available (SMILES).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.