Learn More
Medchemexpress LLC OSMI-4 1mg | 2260791-14-6 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1143611MG
Encompass_Preferred
OSMI-4 is a small-molecule inhibitor of O-GlcNAc transferase (OGT) used as a research reagent; it shows cellular activity with an EC50 of approximately 3 μM and is supplied as a high-purity solid for biochemical and cell-based studies.
  • Low-nanomolar OGT inhibitor with cellular EC50 ≈ 3 μM.
  • Molecular weight 604.09 g/mol; formula C27H26ClN3O7S2.
  • High purity (HPLC) suitable for analytical and biological assays.
  • Available in small research pack sizes (1 mg, 5 mg, 10 mg).
  • Provided with certificate of analysis and safety data sheet for quality and handling information.
Catalog No. 50-003-08125