Learn More
Medchemexpress LLC OSMI-4 50mg | 2260791-14-6 | 50 MG

Supplier: Medchemexpress LLC HY11436150MG
OSMI-4 is a small-molecule inhibitor of O-GlcNAc transferase (OGT) with a cellular EC50 of approximately 3 μM. It is intended for research use in biochemical and cell-based assays.
- Low-nanomolar biochemical potency; cellular EC50 ≈ 3 μM.
- High purity (99.7%), suitable for in vitro studies.
- Soluble in DMSO (≈62.5 mg/mL); ultrasonic assistance recommended.
- Molecular formula C27H26ClN3O7S2; molecular weight 604.09 g·mol⁻¹.
- Available in multiple sizes, including 50 MG.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.