Learn More
Ted Pella Inc Osmium Tetroxide, 4% aqueous, 10 x 2ml
SDP
Supplier:  Ted Pella Inc 18459
Encompass
Solid vaporizes by sublimation. Should be used only under the hood. Affects eyes. Use eye protection. EM Grade, glass ampoules in black paper. No sticky labels. Osmium purity >99.95%. Light sensitive.
Catalog No. NC9402523