Learn More
Medchemexpress LLC Ospemifene | 128607-22-7 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0723100MG
Encompass_Preferred
Ospemifene (FC-1271a) is an orally active and non-estrogenic selective estrogen receptor modulator (SERM) with Ki values of 380 and 410 nM for estrogen receptor α (ERα) and ERβ, respectively. It inhibits caspase-3 activity, neuronal degeneration, prevents bone loss, and increases vaginal weight and vaginal epithelial height. Ospemifene also exhibits anticancer activity against breast cancer.
  • Inhibits caspase-3 activity.
  • Inhibits neuronal degeneration.
  • Prevents bone loss.
  • Increases vaginal weight and vaginal epithelial height.
  • Exhibits anticancer activity against breast cancer.
Catalog No. 50-004-07081