Learn More
Medchemexpress LLC Ossirene | 106566-58-9 | 98.0% | 312.05 | 5 MG
SDP
Supplier:  Medchemexpress LLC 5MGHY101019
Encompass_Preferred
Ossirene is an immunomodulatory tellurium compound and a potent IL-1β inhibitor. It abolishes phosphorylation of STAT3 by inhibiting IL-10 and potently inhibits Caspase-1. This compound is used for autoimmune diseases and certain malignancies.
  • Potent IL-1β inhibitor
  • Inhibits phosphorylation of STAT3 by suppressing IL-10
  • Potently inhibits Caspase-1
  • Used for autoimmune diseases and certain malignancies
  • Sensitizes glioblastoma multiforme (GBM) tumors to paclitaxel
  • Decreases proliferation of B16 melanoma, stomach adenocarcinoma, and human GBM cells
Catalog No. 50-003-65955