Learn More
Lonza Walkersville OsteoImage Mineralization Assay, 500 Test
Supplier:  Lonza Walkersville PA1503
Encompass
The OsteoImage Mineralization Assay is a rapid, fluorescent in vitro assay for assessing bone cell mineralization. The OsteoImage Assay can quantitate in vitro mineralization by osteogenic stem cells, primary osteoblasts, and osteoblast-like cell lines. It is based on specific binding of the fluorescent OsteoImage Staining Reagent to the hydroxyapatite portion of bone-like nodules deposited by cells. Benefits:Delivers qualitative visual fluorescent microscopy or quantitative fluorescent plate reader resultsCan be used with primary osteoblasts, osteoblast stem cells, and osteoblast cell linesMeasures hydroxyapatite, similar to real boneCompleted in less than 90 minutes, without tedious extractionsSensitive enough to detect time dependent increases in mineralization in differentiating cellsScalable for use in 6-well to 96-well plates
Catalog No. NC9952178