Learn More
CPC Scientific H-Leu-Asp-Leu-Asn-Leu-Glu-Leu-Ser-Lys-Phe-Arg-Met-Pro-Gln-Pro-Ser-Ser-Gly-Arg-Glu-Ser-Pro-Arg-His-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific OBAP001B
Encompass
A novel peptide isolated from rat stomach. OBAP positively regulates osteoblast activation and bone formation and therefore has a potential for a therapeutic treatment of osteoporosis and fracture healing. ONE-LETTER SEQUENCE: LDLNLELSKFRMPQPSSGRESPRHMOLECULAR FORMULA: C120H196N38O37S1MOLECULAR WEIGHT:2795.18STORAGE CONDITIONS: -20 5CRESEARCH AREA: OsteoporosisREFERENCES: N.Fukushima et al., Biochem. Biophys. Res. Commun., 400, 157 (2010)
Catalog No. 50-210-9965