Learn More
ATCC Osteocyte Differentiation Tool
Supplier:  ATCC PCS500052
Encompass
The Osteocyte Differentiation Tool is a complete differentiation medium designed to induce osteogenesis in actively proliferating Adipose-derived Mesenchymal Stem Cells (ATCC PCS-500-011) with high efficiency. This product may also be used with Bone Marrow-derived Mesenchymal Stem Cells (ATCC PCS-500-012). The Osteocyte Differentiation Tool provides enough medium for differentiation of ~ 1 million cells when plated at a recommended density of 18,000 viable cells/cm2 in a 6-well tissue culture format.
Catalog No. 50-238-2499