Learn More
ABclonal Technology Osteopontin Mouse mAb
SDP
Supplier:  ABclonal Technology A2108420UL
Encompass_Preferred
The protein encoded by this gene is involved in the attachment of osteoclasts to the mineralized bone matrix The encoded protein is secreted and binds hydroxyapatite with high affinity The osteoclast vitronectin receptor is found in the cell membrane and may be involved in the binding to this protein This protein is also a cytokine that upregulates expression of interferon-gamma and interleukin-12 Several transcript variants encoding different isoforms have been found for this gene
Catalog No. 50-000-07410