Learn More
YSI 3P LifeScience OSTWALD CAPLLRY VISCOMTR 51623
Supplier:  YSI 3P LifeScience 285404236
Encompass
Type No. 516 23, Capillary No. IIc, Constant K = 0.3, calibrated for manual measurements
Catalog No. NC0468560