Learn More
Selleck Chemical LLC OSU-03012 (AR-12) S1106-10mM/1mL
SDP
Supplier:  Selleck Chemical LLC S110610MM/1ML
Encompass_Preferred
OSU-03012 (AR-12) is a potent inhibitor of recombinant PDK-1(phosphoinositide-dependent kinase 1) with IC50 of 5 M in a cell-free assay and 2-fold increase in potency over OSU-02067
Catalog No. 50-310-8449